1,3-Benzenedithiol 95%

Supplier: Thermo Scientific Chemicals
Danger

45610-0010 456100050
TS45610-0010EA 118 USD
TS45610-0010 TS456100050
1,3-Benzenedithiol 95%
1,3-Benzenedithiol
Formula: C₆H₆S₂
Boiling Pt: 128…129 °C (21hPa)
Melting Pt: 24 °C
Density: 1.24 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004842
CAS Number: 626-04-0

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Liquid or solid
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR