Fe(III) meso-Tetra(4-carboxyphenyl)porphine chloride 95%

Supplier: Ambeed
Warning

Synonyms: FeTCPP

A1000418-100MG A1000418-1G A1000418-250MG A1000418-5G
76645-830EA 35.01 USD
76645-830 76645-832 76645-834 77619-466
Fe(III) meso-Tetra(4-carboxyphenyl)porphine chloride 95%
Fe(III) meso-Tetra(4-carboxyphenyl)porphine chloride

Precaución: for Research Use Only

Formula: C₄₈H₂₈ClFeN₄O₈
MW: 880.0559 g/mol
MDL Number: MFCD03425968
CAS Number: 55266-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR