Ethyl 1-(4-nitrophenyl)cyclopropanecarboxylate 95%

Supplier: Ambeed
Warning

A147417-1G A147417-250MG
76647-038EA 105.03 USD
76647-038 76647-040
Ethyl 1-(4-nitrophenyl)cyclopropanecarboxylate 95%
Ethyl 1-(4-nitrophenyl)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.2400 g/mol
MDL Number: MFCD22381468
CAS Number: 1308814-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR