2-(((tert-Butoxycarbonyl)amino)methyl)isonicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A146683-100MG
A146683-1G
A146683-250MG
76647-032EA
172.93
USD
76647-032
76647-034
76647-036
2-(((tert-Butoxycarbonyl)amino)methyl)isonicotinic acid 98%
2-(((tert-Butoxycarbonyl)amino)methyl)isonicotinic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₆N₂O₄ MW: 252.2664 g/mol |
MDL Number:
MFCD11044225 CAS Number: 473924-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...