N-Boc-2-(2-aminoethoxy)acetic Acid 95%

Supplier: Ambeed
Warning

A144692-1G A144692-250MG A144692-25G A144692-5G
76647-024EA 61.54 USD
76647-024 76647-026 76647-028 76647-030
N-Boc-2-(2-aminoethoxy)acetic Acid 95%
N-Boc-2-(2-aminoethoxy)acetic Acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.2350 g/mol
MDL Number: MFCD14155965
CAS Number: 142929-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR