Chlorotris(triphenylphosphine)cobalt(I) 98% + (Co 6.65-6.72%)

Supplier: Ambeed
Warning

A1329295-1G A1329295-250MG A1329295-5G
76657-932EA 15.92 USD
76657-932 76657-934 76657-936
Chlorotris(triphenylphosphine)cobalt(I) 98% + (Co 6.65-6.72%)
Chlorotris(triphenylphosphine)cobalt(I)

Precaución: for Research Use Only

Formula: C₅₄H₄₅ClCoP₃
MW: 881.26 g/mol
MDL Number: MFCD00015864
CAS Number: 26305-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR