4,4',4'',4'''-Methanetetrayltetrabenzaldehyde 98%

Supplier: Ambeed
Warning

A137961-250MG A137961-1G A137961-100MG
76647-000EA 78 USD
76647-000 77566-770 77615-096
4,4',4'',4'''-Methanetetrayltetrabenzaldehyde 98%
4,4',4'',4'''-Methanetetrayltetrabenzaldehyde

Precaución: For research use only.

Formula: C₂₉H₂₀O₄
MW: 432.4667 g/mol
MDL Number: MFCD12911658
CAS Number: 617706-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR