Dimethyl 5-methylpyrimidine-4,6-dicarboxylate 97%

Supplier: Ambeed
Warning

A135838-100MG A135838-1G A135838-250MG
76646-992EA 50.93 USD
76646-992 76646-994 76646-996
Dimethyl 5-methylpyrimidine-4,6-dicarboxylate 97%
Dimethyl 5-methylpyrimidine-4,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.1867 g/mol
MDL Number: MFCD31699630
CAS Number: 2288710-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR