4-(Bromomethyl)-2-methoxypyridine hydrobromide 95%

Supplier: Ambeed
Danger

A134377-1G A134377-250MG A134377-100MG
76646-988EA 96 USD
76646-988 76646-990 77615-074
4-(Bromomethyl)-2-methoxypyridine hydrobromide 95%
4-(Bromomethyl)-2-methoxypyridine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉Br₂NO
MDL Number: MFCD31698061
CAS Number: 2288708-87-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR