1-hexadecyl-3-methylimidazolium hexafluorophosphate 99%

Supplier: Ambeed
Warning

A133281-25G A133281-5G
76646-984EA 21 USD
76646-984 76646-986
1-hexadecyl-3-methylimidazolium hexafluorophosphate 99%
1-hexadecyl-3-methylimidazolium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₂₀H₃₉F₆N₂P
MDL Number: MFCD17014731
CAS Number: 219947-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR