tert-Butyl 4-(1H-indazol-3-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%

Supplier: Ambeed
Warning

A131851-100MG A131851-1G A131851-250MG
76646-954EA 30.77 USD
76646-954 76646-956 76646-958
tert-Butyl 4-(1H-indazol-3-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-(1H-indazol-3-yl)-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁N₃O₂
MW: 299.3675 g/mol
MDL Number: MFCD31699518
CAS Number: 2288708-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR