2-Bromo-1-(m-tolyl)ethan-1-one 97%

Supplier: Ambeed
Danger

A128752-100MG A128752-1G A128752-250MG A128752-5G
76646-938EA 31.83 USD
76646-938 76646-940 76646-942 76646-944
2-Bromo-1-(m-tolyl)ethan-1-one 97%
2-Bromo-1-(m-tolyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.0712 g/mol
MDL Number: MFCD07364269
CAS Number: 51012-64-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR