Methyl 4-(hydrazinylmethyl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

A1329189-100MG A1329189-1G A1329189-250MG
76657-920EA 32.89 USD
76657-920 76657-922 76657-924
Methyl 4-(hydrazinylmethyl)benzoate hydrochloride 97%
Methyl 4-(hydrazinylmethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₂O₂
MW: 216.6647 g/mol
MDL Number: MFCD20485004
CAS Number: 870822-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR