11-Methoxy-11-oxoundecanoic acid 95%

Supplier: Ambeed
Warning

A126747-1G A126747-250MG A126747-10G A126747-25G A126747-5G
76646-918EA 95.49 USD
76646-918 76646-920 77747-906 77747-907 77747-908
11-Methoxy-11-oxoundecanoic acid 95%
11-Methoxy-11-oxoundecanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₄
MW: 230.3007 g/mol
MDL Number: MFCD00673464
CAS Number: 3927-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR