2-Indancarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-1H-indene-2-carboxylic acid, Indan-2-carboxylic acid

A123259-100G A123259-10G A123259-1G A123259-250MG A123259-25G A123259-5G
76646-888EA 387 USD
76646-888 76646-890 76646-892 76646-894 76646-896 76646-898
2-Indancarboxylic acid 95%
2-Indancarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00085095
CAS Number: 25177-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR