4-Methyl-1-(3-nitro-5-(trifluoromethyl)phenyl)-1H-imidazole methanesulfonate 95%

Supplier: Ambeed
Warning

A122528-1G A122528-250MG A122528-5G
76646-882EA 65.78 USD
76646-882 76646-884 76646-886
4-Methyl-1-(3-nitro-5-(trifluoromethyl)phenyl)-1H-imidazole methanesulfonate 95%
4-Methyl-1-(3-nitro-5-(trifluoromethyl)phenyl)-1H-imidazole methanesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃N₃O₅S
MW: 367.3010 g/mol
MDL Number: MFCD01926538
CAS Number: 917391-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR