4-Fluoro-N,N-dimethyl-3-nitroaniline 97%
Supplier: Ambeed
|
Warning
|
A121608-1G
A121608-250MG
76646-878EA
182.48
USD
76646-878
76646-880
4-Fluoro-N,N-dimethyl-3-nitroaniline 97%
4-Fluoro-N,N-dimethyl-3-nitroaniline
Precaución: for Research Use Only
Formula:
C₈H₉FN₂O₂ MW: 184.1677 g/mol |
MDL Number:
MFCD25960351 CAS Number: 18542-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...