(4-Isopropylbenzyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A1329131-1G A1329131-250MG
76657-914EA 151.71 USD
76657-914 76657-916
(4-Isopropylbenzyl)hydrazine hydrochloride 97%
(4-Isopropylbenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂
MW: 200.7084 g/mol
MDL Number: MFCD09696854
CAS Number: 94409-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR