(4-(Hydrazinylmethyl)phenyl)methanol hydrochloride 97%

Supplier: Ambeed
Warning

A1329093-100MG A1329093-250MG
76657-908EA 115.64 USD
76657-908 76657-910
(4-(Hydrazinylmethyl)phenyl)methanol hydrochloride 97%
(4-(Hydrazinylmethyl)phenyl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂O
MW: 188.6546 g/mol
CAS Number: 212914-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR