[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3,5-bis(ethoxycarbonyl)phenyl)boronic acid

A114284-1G A114284-5G
76646-852EA 44.56 USD
76646-852 76646-854
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid 95%
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BO₆
MW: 266.06 g/mol
MDL Number: MFCD11867795
CAS Number: 932378-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR