4-(3-Fluoro-4-nitrophenoxy)-6,7-dimethoxyquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A112854-1G A112854-250MG
76646-846EA 140.04 USD
76646-846 76646-848
4-(3-Fluoro-4-nitrophenoxy)-6,7-dimethoxyquinoline hydrochloride 95%
4-(3-Fluoro-4-nitrophenoxy)-6,7-dimethoxyquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₄ClFN₂O₅
MW: 380.7500 g/mol
CAS Number: 2232877-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR