4,6-Dibromopyrazolo[1,5-a]pyridine-3-carbonitrile 96%

Supplier: Ambeed
Warning

A110189-100MG A110189-1G A110189-250MG
76646-840EA 79.57 USD
76646-840 76646-842 76646-844
4,6-Dibromopyrazolo[1,5-a]pyridine-3-carbonitrile 96%
4,6-Dibromopyrazolo[1,5-a]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₃Br₂N₃
MW: 300.9375 g/mol
MDL Number: MFCD30183519
CAS Number: 2068065-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR