2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile 98%

Supplier: Ambeed
Danger

A109912-1G A109912-250MG A109912-5G
76646-834EA 211.12 USD
76646-834 76646-836 76646-838
2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile 98%
2-(5,6-Difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile

Precaución: for Research Use Only

Formula: C₁₂H₄F₂N₂O
MW: 230.1698 g/mol
MDL Number: MFCD31631729
CAS Number: 2083617-82-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR