2-((4-Chloro-1-guanidinoisoquinoline)-7-sulfonamido)-2-methylpropanoic acid hydrochloride 99+%
Supplier: Ambeed
|
Warning
|
A109441-10MG
A109441-1MG
A109441-25MG
A109441-5MG
76646-826EA
75.33
USD
76646-826
76646-828
76646-830
76646-832
2-((4-Chloro-1-guanidinoisoquinoline)-7-sulfonamido)-2-methylpropanoic acid hydrochloride 99+%
2-((4-Chloro-1-guanidinoisoquinoline)-7-sulfonamido)-2-methylpropanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₇Cl₂N₅O₄S MW: 422.29 g/mol |
MDL Number:
MFCD31558512 CAS Number: 256476-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...