tert-Butyl 4,5-dihydro-2H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate 95%

Supplier: Ambeed
Warning

A108066-100MG A108066-250MG
76646-812EA 100.79 USD
76646-812 76646-814
tert-Butyl 4,5-dihydro-2H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate 95%
tert-Butyl 4,5-dihydro-2H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MW: 223.2716 g/mol
MDL Number: MFCD22381916
CAS Number: 2088956-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR