(S)-2-((tert-Butoxycarbonyl)amino)-5,5,5-trifluoropentanoic acid 98%

Supplier: Ambeed
Warning

A1328918-100MG A1328918-250MG
76657-904EA 27 USD
76657-904 76657-906
(S)-2-((tert-Butoxycarbonyl)amino)-5,5,5-trifluoropentanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-5,5,5-trifluoropentanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆F₃NO₄
MDL Number: MFCD11846980
CAS Number: 453556-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR