(2-Amino-4-(trifluoromethyl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A106136-100MG A106136-1G A106136-250MG
76646-794EA 18 USD
76646-794 76646-796 76646-798
(2-Amino-4-(trifluoromethyl)phenyl)methanol 95%
(2-Amino-4-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MDL Number: MFCD17215842
CAS Number: 186602-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR