Levofloxacin 97%

Supplier: Ambeed
Danger

A105051-100G A105051-100MG A105051-10G A105051-25G A105051-500G A105051-5G
76646-776EA 128 USD
76646-776 76646-778 76646-780 76646-782 76646-784 76646-786
Levofloxacin 97%
Levofloxacin

Precaución: for Research Use Only

Formula: C₁₈H₂₀FN₃O₄
MDL Number: MFCD00865049
CAS Number: 100986-85-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR