2-Bromo-1-(3,4-difluoro-2-hydroxyphenyl)ethan-1-one 95%
Supplier: Ambeed
|
Danger
|
A1045036-1G
A1045036-250MG
76646-772EA
401.03
USD
76646-772
76646-774
2-Bromo-1-(3,4-difluoro-2-hydroxyphenyl)ethan-1-one 95%
2-Bromo-1-(3,4-difluoro-2-hydroxyphenyl)ethan-1-one
Precaución: for Research Use Only
Formula:
C₈H₅BrF₂O₂ MW: 251.0249 g/mol |
MDL Number:
MFCD28789107 CAS Number: 1621375-69-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...