5-Bromo-1-(cyclopropylmethyl)-1,2-dihydropyridin-2-one 97%

Supplier: Ambeed
Danger

A1039427-100MG A1039427-1G A1039427-250MG
76646-766EA 29 USD
76646-766 76646-768 76646-770
5-Bromo-1-(cyclopropylmethyl)-1,2-dihydropyridin-2-one 97%
5-Bromo-1-(cyclopropylmethyl)-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MDL Number: MFCD21746683
CAS Number: 1114573-41-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR