6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1026186-1G A1026186-50MG
76646-750EA 310 USD
76646-750 76646-752
6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-carboxylic acid 95%
6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MDL Number: MFCD20620912
CAS Number: 19250-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR