(2S)-2-(4-Fluorophenyl)-2-hydroxyacetic acid 95%
Supplier: Ambeed
|
Warning
|
A1020684-100MG
A1020684-1G
76646-746EA
28
USD
76646-746
77619-678
(2S)-2-(4-Fluorophenyl)-2-hydroxyacetic acid 95%
(2S)-2-(4-Fluorophenyl)-2-hydroxyacetic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD20643876 CAS Number: 52923-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...