2,2'-Difluorobenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: Bis(2-fluorophenyl)methanone

A1014078-250MG A1014078-1G A1014078-100MG
76646-734EA 194.15 USD
76646-734 77570-102 77619-668
2,2'-Difluorobenzophenone 95%
2,2'-Difluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O
MW: 218.1988 g/mol
MDL Number: MFCD12025148
CAS Number: 342-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR