2,5-Dioxopyrrolidin-1-yl 3-mercaptopropanoate 95%

Supplier: Ambeed
Danger

A1327929-100MG A1327929-1G A1327929-250MG
76657-900EA 29 USD
76657-900 77566-588 77621-544
2,5-Dioxopyrrolidin-1-yl 3-mercaptopropanoate 95%
2,5-Dioxopyrrolidin-1-yl 3-mercaptopropanoate

Precaución: For research use only.

Formula: C₇H₉NO₄S
MDL Number: MFCD28964889
CAS Number: 117235-10-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR