1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one 95%
Supplier: Ambeed
|
Warning
|
A1011058-1G
A1011058-250MG
76646-716EA
176.11
USD
76646-716
76646-718
1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one 95%
1-[4-(Dimethylamino)-2-hydroxyphenyl]ethan-1-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂ MW: 179.2157 g/mol |
MDL Number:
MFCD11870193 CAS Number: 107070-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...