Methyl 5-bromo-2-(chlorosulfonyl)benzoate 95%

Supplier: Ambeed
Danger

A1010062-100MG A1010062-1G A1010062-250MG A1010062-5G
76646-708EA 59 USD
76646-708 76646-710 76646-712 77619-656
Methyl 5-bromo-2-(chlorosulfonyl)benzoate 95%
Methyl 5-bromo-2-(chlorosulfonyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₄S
MDL Number: MFCD04638740
CAS Number: 736948-72-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR