1-Propyl-2-thioxodihydropyrimidine-4,6(1H,5H)-dione 95%
Supplier: Ambeed
|
Warning
|
A1008947-100MG
A1008947-1G
76646-696EA
26.53
USD
76646-696
76646-698
1-Propyl-2-thioxodihydropyrimidine-4,6(1H,5H)-dione 95%
1-Propyl-2-thioxodihydropyrimidine-4,6(1H,5H)-dione
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₂S MW: 186.2315 g/mol |
MDL Number:
MFCD03976141 CAS Number: 65959-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...