Tabersonine hydrochloride 98%

Supplier: Ambeed
Warning

A1006835-100G A1006835-25G A1006835-5G
76646-692EA 154 USD
76646-692 77619-652 77619-654
Tabersonine hydrochloride 98%
Tabersonine hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₂
MDL Number: MFCD02258999
CAS Number: 29479-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR