(S)-5-Oxopyrrolidine-2-carboxylic acid compound with (1S,2S,3S,4R,5S)-5-(4-chloro-3-(4-ethoxybenzyl)phenyl)-1-(hydroxymethyl)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol (1:1) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ertugliflozin L-pyroglutamic acid
A1006406-10MG
A1006406-1MG
A1006406-5MG
A1006406-100MG
A1006406-250MG
76646-676EA
19.1
USD
76646-676
76646-678
76646-680
77619-640
77619-642
(S)-5-Oxopyrrolidine-2-carboxylic acid compound with (1S,2S,3S,4R,5S)-5-(4-chloro-3-(4-ethoxybenzyl)phenyl)-1-(hydroxymethyl)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol (1:1) 99%
(S)-5-Oxopyrrolidine-2-carboxylic acid compound with (1S,2S,3S,4R,5S)-5-(4-chloro-3-(4-ethoxybenzyl)phenyl)-1-(hydroxymethyl)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol (1:1)
Precaución: for Research Use Only
Formula:
C₂₇H₃₂ClNO₁₀ MW: 566 g/mol |
MDL Number:
MFCD27956909 CAS Number: 1210344-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...