(2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2Sp)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene

A1006373-100MG A1006373-250MG A1006373-50MG
76646-672EA 107.16 USD
76646-672 76646-674 77619-638
(2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene ≥97%, ee 99%
(2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₃₆H₃₆FeO₂P₂
MW: 618.4600 g/mol
MDL Number: MFCD06658128
CAS Number: 649559-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR