Tetrakisacetonitrile copper(I) triflate 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(acetonitrile)copper trifluoromethanesulphonate, Tetrakis(acetonitrile)copper(I) Trifluoromethanesulphonate

A1006248-100MG A1006248-1G A1006248-250MG A1006248-5G A1006248-25G
76646-664EA 5 USD
76646-664 76646-666 76646-668 76646-670 77782-820
Tetrakisacetonitrile copper(I) triflate 98%
Tetrakisacetonitrile copper(I) triflate

Precaución: for Research Use Only

Formula: Cu(NCCH₃)₄·CF₃SO₃
MW: 376.821 g/mol
MDL Number: MFCD10566992
CAS Number: 58452-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR