1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-((1S,2S,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methyl)thiourea 98%

Supplier: Ambeed
Danger

A1006185-100MG A1006185-250MG A1006185-5G
76646-654EA 55 USD
76646-654 77782-815 77782-816
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-((1S,2S,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methyl)thiourea 98%
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((S)-((1S,2S,4S,5R)-5-ethylquinuclidin-2-yl)(6-methoxyquinolin-4-yl)methyl)thiourea

Precaución: for Research Use Only

Formula: C₂₉H₃₀F₆N₄OS
MDL Number: MFCD16875669
CAS Number: 852913-19-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR