(R)-5-Benzyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1006163-100MG A1006163-250MG
76646-650EA 132.62 USD
76646-650 76646-652
(R)-5-Benzyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
(R)-5-Benzyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₄BF₄N₃
MW: 405.2400 g/mol
CAS Number: 1469801-67-9
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR