(S)-5-Isopropyl-2-mesityl-6,8-dihydro-5H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1006162-250MG A1006162-100MG
76646-648EA 143 USD
76646-648 77619-618
(S)-5-Isopropyl-2-mesityl-6,8-dihydro-5H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%
(S)-5-Isopropyl-2-mesityl-6,8-dihydro-5H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₇H₂₄BF₄N₃O
MW: 373.1966 g/mol
MDL Number: MFCD32655223
CAS Number: 1631733-83-9
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR