(11bR)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%

Supplier: Ambeed
Warning

A1006137-100MG A1006137-1G A1006137-250MG
76646-626EA 113.52 USD
76646-626 76646-628 76646-630
(11bR)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bR)-2,6-Bis(4-chlorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₃₂H₁₉Cl₂O₄P
MW: 569.3706 g/mol
CAS Number: 922711-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR