(11bR)-N-(2,6-Bis(4-nitrophenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1006136-100MG A1006136-1G A1006136-250MG A1006136-50MG
76646-618EA 95 USD
76646-618 76646-620 76646-622 76646-624
(11bR)-N-(2,6-Bis(4-nitrophenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bR)-N-(2,6-Bis(4-nitrophenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₃₃H₁₉F₃N₃O₉PS
CAS Number: 1010799-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR