(11bR)-N-(2,6-Di(naphthalen-2-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
Supplier: Ambeed
|
Warning
|
A1006127-100MG
A1006127-1G
A1006127-250MG
A1006127-50MG
76646-594EA
49
USD
76646-594
76646-596
76646-598
76646-600
(11bR)-N-(2,6-Di(naphthalen-2-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bR)-N-(2,6-Di(naphthalen-2-yl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide
Precaución: for Research Use Only
Formula:
C₄₁H₃₃F₃NO₅PS |
MDL Number:
MFCD33548845 CAS Number: 1261302-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...