(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1006125-100MG A1006125-1G A1006125-250MG A1006125-50MG
76646-586EA 197.33 USD
76646-586 76646-588 76646-590 76646-592
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxido-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₃₇H₂₅F₁₅NO₅PS
MW: 911.6129 g/mol
CAS Number: 1261302-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR