(11bS)-2,6-Dimethyl-N,N-bis(1-phenylethyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥97%, ee 99%

Supplier: Ambeed
Warning

A1006119-1G A1006119-250MG
76646-576EA 618.51 USD
76646-576 76646-578
(11bS)-2,6-Dimethyl-N,N-bis(1-phenylethyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥97%, ee 99%
(11bS)-2,6-Dimethyl-N,N-bis(1-phenylethyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

Precaución: for Research Use Only

Formula: C₃₈H₃₄NO₂P
MW: 567.6500 g/mol
MDL Number: MFCD16876573
CAS Number: 201732-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR