(11bR)-(2R,5R)-1-(Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-2,5-diphenylpyrrolidine 98%
Supplier: Ambeed
|
Warning
|
A1006117-250MG
A1006117-100MG
A1006117-1G
76646-574EA
347.98
USD
76646-574
77619-606
77619-608
(11bR)-(2R,5R)-1-(Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-2,5-diphenylpyrrolidine 98%
(11bR)-(2R,5R)-1-(Dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-2,5-diphenylpyrrolidine
Precaución: for Research Use Only
Formula:
C₃₆H₂₈NO₂P MW: 537.5800 g/mol |
MDL Number:
MFCD16621390 CAS Number: 444667-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...